siRNA Details
| Gene Symbol: Nags | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145829.2 | NP_665828.1 | 6 | 1344 | ||
| GenBank mRNA | AF462069.1 | - | 6 | 1300 | ||
| AJ489814.1 | - | 6 | 1270 | |||
| AK075765.1 | - | 5 | 990 | |||
| BC036161.1 | - | 3 | 1995 | |||
| BC057990.1 | - | 6 | 1308 | 5100021 | ||

Nags
Genomic Map