siRNA Details
| Gene Symbol: Abca9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147220.2 | NP_671753.2 | 23 | 3372 | ||
| GenBank mRNA | AF491299.1 | - | 23 | 3372 | ||
| AK029256.1 | - | 9 | 1161 | |||
| AK085555.1 | - | 23 | 3280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca9| Gene Symbol: Abca9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147220.2 | NP_671753.2 | 23 | 3372 | ||
| GenBank mRNA | AF491299.1 | - | 23 | 3372 | ||
| AK029256.1 | - | 9 | 1161 | |||
| AK085555.1 | - | 23 | 3280 | |||