siRNA Details
| Gene Symbol: Cd300lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199221.2 | NP_954691.2 | 2 | 483 | ||
| GenBank mRNA | AB300703.1 | - | 2 | 345 | ||
| AK154359.1 | - | 2 | 484 | |||
| AK169466.1 | - | 2 | 233 | |||
| AK170613.1 | - | 2 | 455 | |||
| AY457053.1 | - | 2 | 345 | |||
| AY996128.1 | - | 2 | 384 | |||
| BC107351.1 | - | 2 | 345 | |||
| BC107352.1 | - | 2 | 121 | |||

Cd300lb
Genomic Map