siRNA Details
| Gene Symbol: Cd300lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199221.2 | NP_954691.2 | 3 | 521 | ||
| GenBank mRNA | AB300703.1 | - | 3 | 383 | ||
| AK154359.1 | - | 3 | 522 | |||
| AK169466.1 | - | 3 | 271 | |||
| AK170613.1 | - | 3 | 493 | |||
| AY457053.1 | - | 3 | 383 | |||
| AY996128.1 | - | 3 | 422 | |||
| BC107351.1 | - | 3 | 383 | |||
| BC107352.1 | - | 3 | 159 | |||

Cd300lb
Genomic Map