siRNA Details
| Gene Symbol: Inmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009349.3 | NP_033375.1 | 1 | 60 | ||
| GenBank mRNA | AK002281.1 | - | 1 | 108 | ||
| AK013010.1 | - | 1 | 97 | |||
| BC013518.1 | - | 1 | 86 | 4236077 | ||
| M88694.1 | - | 1 | 61 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inmt| Gene Symbol: Inmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009349.3 | NP_033375.1 | 1 | 60 | ||
| GenBank mRNA | AK002281.1 | - | 1 | 108 | ||
| AK013010.1 | - | 1 | 97 | |||
| BC013518.1 | - | 1 | 86 | 4236077 | ||
| M88694.1 | - | 1 | 61 | |||