siRNA Details
| Gene Symbol: Tesk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011571.3 | NP_035701.3 | 8 | 1986 | ||
| GenBank mRNA | AB003494.1 | - | 8 | 2324 | ||
| AK144302.1 | - | 8 | 1569 | |||
| BC099699.1 | - | 8 | 1986 | 6821934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tesk1| Gene Symbol: Tesk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011571.3 | NP_035701.3 | 8 | 1986 | ||
| GenBank mRNA | AB003494.1 | - | 8 | 2324 | ||
| AK144302.1 | - | 8 | 1569 | |||
| BC099699.1 | - | 8 | 1986 | 6821934 | ||