siRNA Details
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161365.1 | NP_001154837.1 | 4 | 512 | ||
NM_177620.4 | NP_808288.2 | 4 | 623 | |||
GenBank mRNA | AK032205.1 | - | 4 | 624 | ||
AK133988.1 | - | 4 | 531 | |||
AK154506.1 | - | 4 | 650 | |||
AK154522.1 | - | 4 | 427 | |||
AK154963.1 | - | 4 | 513 | |||
AK166196.1 | - | 4 | 469 | |||
AK171301.1 | - | 4 | 436 | |||
AK171311.1 | - | 4 | 427 | |||
AY265356.1 | - | 4 | 609 | |||
BC137907.1 | - | 4 | 551 | 8860926 |
