siRNA Details
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161365.1 | NP_001154837.1 | 6 | 789 | ||
NM_177620.4 | NP_808288.2 | 6 | 900 | |||
GenBank mRNA | AK032205.1 | - | 6 | 901 | ||
AK154506.1 | - | 6 | 925 | |||
AK154522.1 | - | 6 | 704 | |||
AK154963.1 | - | 6 | 790 | |||
AK166196.1 | - | 6 | 746 | |||
AK171301.1 | - | 6 | 713 | |||
AK171311.1 | - | 6 | 705 | |||
AY265356.1 | - | 6 | 886 | |||
BC137907.1 | - | 6 | 828 | 8860926 |
