siRNA Details
| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146039.3 | NP_666151.3 | 7 | 1316 | ||
| GenBank mRNA | AK046399.1 | - | 7 | 1278 | ||
| AK052482.1 | - | 7 | 1280 | |||
| AK144138.1 | - | 7 | 1316 | |||
| BC138916.1 | - | 7 | 1249 | |||

Wdr60
Genomic Map