siRNA Details
| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146039.3 | NP_666151.3 | 5 | 961 | ||
| GenBank mRNA | AK046399.1 | - | 5 | 923 | ||
| AK052482.1 | - | 5 | 925 | |||
| AK144138.1 | - | 5 | 961 | |||
| BC138916.1 | - | 5 | 894 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr60| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146039.3 | NP_666151.3 | 5 | 961 | ||
| GenBank mRNA | AK046399.1 | - | 5 | 923 | ||
| AK052482.1 | - | 5 | 925 | |||
| AK144138.1 | - | 5 | 961 | |||
| BC138916.1 | - | 5 | 894 | |||