siRNA Details
Gene Symbol: Zfp455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048204.1 | NP_001041669.1 | 4 | 1042 | ||
GenBank mRNA | AK028336.1 | - | 3 | 887 | ||
BC010651.1 | - | 1 | 489 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048204.1 | NP_001041669.1 | 4 | 1042 | ||
GenBank mRNA | AK028336.1 | - | 3 | 887 | ||
BC010651.1 | - | 1 | 489 |