siRNA Details
| Gene Symbol: Khnyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027143.2 | NP_081419.1 | 7 | 1889 | ||
| GenBank mRNA | AK032037.1 | - | 7 | 1861 | ||
| AK033704.1 | - | 7 | 1890 | |||
| AK047054.1 | - | 7 | 2435 | |||
| AK047955.1 | - | 7 | 1858 | |||
| AK122247.1 | - | 7 | 1901 | |||
| AK136301.1 | - | 7 | 1917 | |||
| BC116260.1 | - | 7 | 1876 | 40093805 | ||
| BC116261.1 | - | 7 | 1876 | 40093806 | ||

Khnyn
Genomic Map