siRNA Details
| Gene Symbol: Shisa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145463.5 | NP_663438.1 | 2 | 1391 | ||
| GenBank mRNA | AB280737.1 | - | 2 | 1145 | ||
| AK034905.1 | - | 2 | 1366 | |||
| AK087512.1 | - | 2 | 1361 | |||
| BC024118.1 | - | 2 | 827 | 5068886 | ||
| BC057640.1 | - | 2 | 955 | 4018213 | ||
| DQ342342.1 | - | 2 | 605 | |||

Shisa2
Genomic Map