siRNA Details
| Gene Symbol: Trex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012236.1 | NP_001012236.1 | 2 | 886 | ||
| NM_011637.6 | NP_035767.4 | 2 | 900 | |||
| GenBank mRNA | AF151106.3 | - | 2 | 886 | ||
| AF319574.1 | - | 1 | 1600 | |||
| AK009899.1 | - | 2 | 887 | |||
| AK087832.1 | - | 2 | 887 | |||
| AK171916.1 | - | 2 | 875 | |||
| AU041952.1 | - | 1 | 171 | |||
| BC011133.1 | - | 2 | 907 | 4206812 | ||
| CO041929.1 | - | 1 | 183 | |||

Trex1
Genomic Map