siRNA Details
| Gene Symbol: Trpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009428.2 | NP_033454.1 | 6 | 2008 | ||
| GenBank mRNA | AF029983.1 | - | 6 | 1663 | ||
| AF060107.1 | - | 6 | 2007 | |||
| AJ006204.1 | - | 5 | 1605 | |||
| AK038531.1 | - | 6 | 1832 | |||
| AK047867.1 | - | 4 | 1029 | |||
| BC112972.1 | - | 6 | 1826 | 40049027 | ||

Trpc5
Genomic Map