siRNA Details
| Gene Symbol: Tulp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021478.1 | NP_067453.1 | 11, 12 | 1139 | ||
| GenBank mRNA | AF085681.1 | - | 11, 12 | 1187 | ||
| AF105711.1 | - | 11, 12 | 1187 | |||
| AK053880.1 | - | 5, 6 | 642 | |||
| AK164621.1 | - | 1, 2 | 143 | |||
| BC014820.1 | - | 11, 12 | 1145 | |||
| BC029222.1 | - | 11, 12 | 1178 | 4504473 | ||

Tulp1
Genomic Map