siRNA Details
| Gene Symbol: Tulp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021478.1 | NP_067453.1 | 12, 13 | 1248 | ||
| GenBank mRNA | AF085681.1 | - | 12, 13 | 1296 | ||
| AF105711.1 | - | 12, 13 | 1296 | |||
| AK053880.1 | - | 6, 7 | 751 | |||
| AK164621.1 | - | 2, 3 | 252 | |||
| BC014820.1 | - | 12, 13 | 1254 | |||
| BC029222.1 | - | 12, 13 | 1287 | 4504473 | ||

Tulp1
Genomic Map