siRNA Details
| Gene Symbol: Uchl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011670.2 | NP_035800.2 | 9 | 768 | ||
| GenBank mRNA | AB025313.1 | - | 9 | 619 | ||
| AF172334.1 | - | 9 | 607 | |||
| AK004066.1 | - | 9 | 645 | |||
| AK013729.1 | - | 9 | 648 | |||
| AK170728.1 | - | 7 | 606 | |||
| BC039177.1 | - | 9 | 634 | 4506229 | ||

Uchl1
Genomic Map