siRNA Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011672.4 | NP_035802.3 | 10 | 872 | ||
| GenBank mRNA | AK010461.1 | - | 10 | 841 | ||
| AK088823.1 | - | 10 | 841 | |||
| AK147138.1 | - | 10 | 855 | |||
| AK169062.1 | - | 10 | 854 | |||
| BC006630.1 | - | 10 | 829 | 3708320 | ||
| U64445.1 | - | 10 | 857 | |||

Ufd1l
Genomic Map