siRNA Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136086.2 | NP_001129558.1 | 3 | 420 | ||
| NM_001291455.1 | NP_001278384.1 | 3 | 670 | |||
| NM_009468.5 | NP_033494.1 | 3 | 601 | |||
| GenBank mRNA | AF501324.1 | - | 3 | 621 | ||
| AK077000.1 | - | 3 | 411 | |||
| AK133040.1 | - | 3 | 420 | |||
| AK149024.1 | - | 3 | 439 | |||
| AK161509.1 | - | 3 | 342 | |||
| BC023003.1 | - | 3 | 285 | 5367381 | ||
| X87817.1 | - | 3 | 561 | |||

Dpysl3
Genomic Map