siRNA Details
| Gene Symbol: Vmn2r89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009486.3 | NP_033512.1 | 2 | 499 | ||
| NM_009487.1 | NP_033513.1 | 2 | 499 | |||
| GenBank mRNA | AF011420.1 | - | 2 | 498 | ||
| AF011421.1 | - | 2 | 499 | |||
| BC145855.1 | - | 2 | 460 | 40131158 | ||
More Information
Set Membership: |
|

Vmn2r89
Genomic Map