siRNA Details
| Gene Symbol: Ugt3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144845.3 | NP_659094.1 | 4 | 481 | ||
| GenBank mRNA | AK041045.1 | - | 4 | 466 | ||
| AK050128.1 | - | 4 | 482 | |||
| AK052644.1 | - | 4 | 491 | |||
| AK143815.1 | - | 4 | 496 | |||
| BC022134.1 | - | 4 | 466 | 5098754 | ||
| BC034837.1 | - | 4 | 462 | 5134131 | ||

Ugt3a2
Genomic Map