siRNA Details
| Gene Symbol: Fam49b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144846.5 | NP_659095.1 | 4 | 455 | ||
| GenBank mRNA | AK028062.1 | - | 4 | 456 | ||
| AK075613.1 | - | 5 | 400 | |||
| AK205511.1 | - | 2 | 113 | |||
| AK205521.1 | - | 3 | 204 | |||
| BC011343.1 | - | 4 | 273 | 4012217 | ||

Fam49b
Genomic Map