siRNA Details
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016852.2 | NP_058548.1 | 2, 3 | 209 | ||
GenBank mRNA | AK004824.1 | - | 2, 3 | 211 | ||
AK043037.1 | - | 2, 3 | 183 | |||
AK146572.1 | - | 2, 3 | 211 | |||
AK150018.1 | - | 2, 3 | 184 | |||
AK151368.1 | - | 2, 3 | 201 | |||
AK162810.1 | - | 2, 3 | 184 | |||
AK170317.1 | - | 2, 3 | 210 | |||
AK170474.1 | - | 2, 3 | 212 | |||
BC031908.1 | - | 2, 3 | 172 | 5365118 | ||
BC055058.1 | - | 2, 3 | 229 | 6529458 | ||
U40826.1 | - | 2, 3 | 209 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK044015.1 | - | 199 | Chr. - - [-] on Build |
