siRNA Details
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016852.2 | NP_058548.1 | 7 | 732 | ||
GenBank mRNA | AK043037.1 | - | 7 | 706 | ||
AK146572.1 | - | 7 | 734 | |||
AK150018.1 | - | 7 | 707 | |||
AK151368.1 | - | 7 | 724 | |||
AK170317.1 | - | 7 | 733 | |||
AK170474.1 | - | 7 | 735 | |||
BC031908.1 | - | 7 | 695 | 5365118 | ||
BC055058.1 | - | 7 | 752 | 6529458 | ||
U40826.1 | - | 7 | 732 |
