siRNA Details
| Gene Symbol: Fmnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011711.1 | NP_035841.1 | 16 | 1916 | ||
| GenBank mRNA | AK129473.1 | - | 16 | 1835 | ||
| AK139711.1 | - | 16 | 1900 | |||
| AK143308.1 | - | 16 | 1816 | |||
| AK155331.1 | - | 16 | 1933 | |||
| AK163361.1 | - | 16 | 1903 | |||
| AK170744.1 | - | 15 | 1746 | |||
| AK195327.1 | - | 1 | 44 | |||
| BC023797.1 | - | 15 | 1636 | |||
| BC085094.1 | - | 16 | 1811 | |||
| BC131961.1 | - | 16 | 1792 | 40130238 | ||
| U92453.1 | - | 5 | 590 | |||

Fmnl3
Genomic Map