siRNA Details
| Gene Symbol: Wbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011712.2 | NP_035842.1 | 3 | 443 | ||
| GenBank mRNA | AK002214.1 | - | 3 | 397 | ||
| AK213578.1 | - | 1 | 50 | |||
| AY459189.1 | - | 3 | 442 | |||
| BC007478.1 | - | 3 | 340 | 3490650 | ||
| BC020289.1 | - | 3 | 368 | |||
| BU511227.1 | - | 3 | 474 | |||
| U92454.1 | - | 1 | 338 | |||

Wbp5
Genomic Map