siRNA Details
| Gene Symbol: Rnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172612.3 | NP_766200.1 | 5 | 753 | ||
| GenBank mRNA | AK043639.1 | - | 5 | 754 | ||
| AK161870.1 | - | 5 | 750 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnd1| Gene Symbol: Rnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172612.3 | NP_766200.1 | 5 | 753 | ||
| GenBank mRNA | AK043639.1 | - | 5 | 754 | ||
| AK161870.1 | - | 5 | 750 | |||