siRNA Details
Gene Symbol: Rnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172612.3 | NP_766200.1 | 5 | 589 | ||
GenBank mRNA | AK043639.1 | - | 5 | 590 | ||
AK161870.1 | - | 5 | 586 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172612.3 | NP_766200.1 | 5 | 589 | ||
GenBank mRNA | AK043639.1 | - | 5 | 590 | ||
AK161870.1 | - | 5 | 586 |