siRNA Details
| Gene Symbol: Xk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023500.2 | NP_075989.1 | 3 | 1350 | ||
| GenBank mRNA | AF155511.1 | - | 3 | 1285 | ||
| AK007734.1 | - | 3 | 1350 | |||
| AY534248.1 | - | 3 | 1330 | |||
| BC027097.1 | - | 3 | 1377 | |||
| BC125618.1 | - | 3 | 1227 | 40130133 | ||
| BC125620.1 | - | 3 | 1227 | 40130135 | ||

Xk
Genomic Map