siRNA Details
Gene Symbol: Dpp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172624.3 | NP_766212.2 | 5 | 728 | ||
GenBank mRNA | AK039652.1 | - | 5 | 746 | ||
AK050021.1 | - | 5 | 670 | |||
AK050023.1 | - | 5 | 730 | |||
AK078301.1 | - | 5 | 676 | |||
AK131178.1 | - | 2 | 2591 | |||
AK195502.1 | - | 1 | 55 | |||
BC057631.1 | - | 5 | 727 | 4191886 |
