siRNA Details
| Gene Symbol: Gm94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033280.2 | NP_001028452.1 | 1 | 48 | ||
| GenBank mRNA | AK131907.1 | - | 1 | 49 | ||
| AK131908.1 | - | 1 | 49 | |||
| BC147098.1 | - | 1 | 37 | 9056146 | ||
| BC147099.1 | - | 1 | 37 | 9056134 | ||
| BC171995.1 | - | 1 | 37 | 9054281 | ||

Gm94
Genomic Map