siRNA Details
| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178686.3 | NP_848801.2 | 20 | 2682 | ||
| GenBank mRNA | AK038916.1 | - | 20 | 2744 | ||
| AK080073.1 | - | 1 | 559 | |||
| AK089236.1 | - | 20 | 2690 | |||
| AK089242.1 | - | 20 | 2690 | |||
| BC046493.1 | - | 12 | 1548 | |||
| BC053439.1 | - | 20 | 2682 | 6328829 | ||

Cep120
Genomic Map