siRNA Details
| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178686.3 | NP_848801.2 | 19 | 2647 | ||
| GenBank mRNA | AK038916.1 | - | 19 | 2709 | ||
| BC046493.1 | - | 11 | 1513 | |||
| BC053439.1 | - | 19 | 2647 | 6328829 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep120| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178686.3 | NP_848801.2 | 19 | 2647 | ||
| GenBank mRNA | AK038916.1 | - | 19 | 2709 | ||
| BC046493.1 | - | 11 | 1513 | |||
| BC053439.1 | - | 19 | 2647 | 6328829 | ||