siRNA Details
| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178686.3 | NP_848801.2 | 15, 16 | 2199 | ||
| GenBank mRNA | AK038916.1 | - | 15, 16 | 2261 | ||
| AK044808.1 | - | 14, 15 | 3264 | |||
| AK089236.1 | - | 15, 16 | 2206 | |||
| AK089242.1 | - | 15, 16 | 2206 | |||
| BC046493.1 | - | 7, 8 | 1065 | |||
| BC053439.1 | - | 15, 16 | 2199 | 6328829 | ||

Cep120
Genomic Map