siRNA Details
| Gene Symbol: Xpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009531.2 | NP_033557.2 | 9 | 1746 | ||
| GenBank mRNA | AB071144.1 | - | 9 | 1683 | ||
| AK004713.1 | - | 9 | 1686 | |||
| AK028595.1 | - | 9 | 1684 | |||
| AK166981.1 | - | 9 | 1681 | |||
| AK170975.1 | - | 9 | 1678 | |||
| BC052725.1 | - | 9 | 1708 | 6812198 | ||
| U27398.1 | - | 9 | 1657 | |||
| U40005.1 | - | 8 | 1485 | |||

Xpc
Genomic Map