siRNA Details
| Gene Symbol: Glis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054020.1 | - | 4 | 1619 | ||
| AY220846.1 | - | 2 | 664 | |||
| BC066143.1 | - | 3 | 999 | |||
| BC129906.1 | - | 2 | 642 | |||
| BC129907.1 | - | 2 | 642 | |||
| DQ871027.1 | - | 4 | 1619 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glis3| Gene Symbol: Glis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054020.1 | - | 4 | 1619 | ||
| AY220846.1 | - | 2 | 664 | |||
| BC066143.1 | - | 3 | 999 | |||
| BC129906.1 | - | 2 | 642 | |||
| BC129907.1 | - | 2 | 642 | |||
| DQ871027.1 | - | 4 | 1619 | |||