siRNA Details
| Gene Symbol: Ywhag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018871.3 | NP_061359.2 | 2 | 865 | ||
| GenBank mRNA | AB073970.1 | - | 2 | 812 | ||
| AF058799.1 | - | 2 | 815 | |||
| AK088847.1 | - | 2 | 778 | |||
| AK148618.1 | - | 2 | 824 | |||
| AK153307.1 | - | 2 | 824 | |||
| AK164356.1 | - | 2 | 816 | |||
| BC008129.1 | - | 2 | 748 | 3498633 | ||
| CT010208.1 | - | 2 | 633 | |||

Ywhag
Genomic Map