siRNA Details
| Gene Symbol: Zap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289612.1 | NP_001276541.1 | 1 | 144 | ||
| NM_001289765.1 | NP_001276694.1 | 7, 8 | 1091 | |||
| NM_001289766.1 | NP_001276695.1 | 7, 8 | 1056 | |||
| NM_009539.3 | NP_033565.2 | 7, 8 | 1048 | |||
| GenBank mRNA | AB083210.1 | - | 1 | 147 | ||
| AK039883.1 | - | 7, 8 | 985 | |||
| AK089056.1 | - | 7, 8 | 1021 | |||
| BC029727.1 | - | 7, 8 | 1006 | 4925739 | ||
| U04379.1 | - | 7, 8 | 1002 | |||
| U77667.1 | - | 6, 7 | 894 | |||

Zap70
Genomic Map