siRNA Details
| Gene Symbol: Ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144880.4 | NP_659129.2 | 5, 6 | 1334 | ||
| GenBank mRNA | AK145397.1 | - | 6, 7 | 720 | ||
| BC059026.1 | - | 5, 6 | 1334 | 6810754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r5a| Gene Symbol: Ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144880.4 | NP_659129.2 | 5, 6 | 1334 | ||
| GenBank mRNA | AK145397.1 | - | 6, 7 | 720 | ||
| BC059026.1 | - | 5, 6 | 1334 | 6810754 | ||