siRNA Details
Gene Symbol: Zfp92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009566.4 | NP_033592.2 | 6 | 566 | ||
GenBank mRNA | AK141522.1 | - | 6 | 567 | ||
BC096436.1 | - | 5 | 617 | 30478238 | ||
U47104.1 | - | 6 | 586 |
