siRNA Details
| Gene Symbol: Proser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159657.1 | NP_001153129.1 | 3 | 687 | ||
| NM_144883.4 | NP_659132.2 | 3 | 541 | |||
| GenBank mRNA | AK077380.1 | - | 3 | 517 | ||
| AK132370.1 | - | 491 | ||||
| BC023193.1 | - | 2 | 400 | 4946741 | ||
| BC025867.1 | - | 4 | 648 | 5052194 | ||
| BC029968.1 | - | 3 | 504 | |||

Proser2
Genomic Map