siRNA Details
Gene Symbol: Proser2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159657.1 | NP_001153129.1 | 4 | 1176 | ||
NM_144883.4 | NP_659132.2 | 4 | 1030 | |||
GenBank mRNA | AK077380.1 | - | 4 | 1006 | ||
AK080430.1 | - | 1 | 441 | |||
AK132370.1 | - | 980 | ||||
BC023193.1 | - | 3 | 889 | 4946741 | ||
BC025867.1 | - | 5 | 1137 | 5052194 | ||
BC026904.1 | - | 1 | 100 | |||
BC029968.1 | - | 4 | 993 |
