siRNA Details
Gene Symbol: Ppapdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145521.3 | NP_663496.1 | 1 | 421 | ||
GenBank mRNA | AK052883.1 | - | 1 | 422 | ||
AK134638.1 | - | 1 | 545 | |||
BC016136.1 | - | 1 | 352 | 4910887 |
