siRNA Details
| Gene Symbol: Zic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009576.2 | NP_033602.2 | 3 | 776 | ||
| GenBank mRNA | AK028305.1 | - | 1 | 579 | ||
| AK035908.1 | - | 1 | 580 | |||
| AK046913.1 | - | 3 | 777 | |||
| AK163477.1 | - | 3 | 1850 | |||
| D78174.1 | - | 1 | 1029 | |||

Zic4
Genomic Map