siRNA Details
| Gene Symbol: Xkr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011732.1 | NP_001011732.1 | 2 | 817 | ||
| GenBank mRNA | AY534253.1 | - | 2 | 817 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr7| Gene Symbol: Xkr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011732.1 | NP_001011732.1 | 2 | 817 | ||
| GenBank mRNA | AY534253.1 | - | 2 | 817 | ||