siRNA Details
| Gene Symbol: Fam151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146149.1 | NP_666261.1 | 2, 3 | 323 | ||
| GenBank mRNA | AY168619.1 | - | 2, 3 | 323 | ||
| BC024426.1 | - | 2, 3 | 323 | 4971419 | ||
| BC024430.1 | - | 2, 3 | 323 | 4972368 | ||
| BC026682.1 | - | 2, 3 | 332 | 5065146 | ||

Fam151a
Genomic Map