siRNA Details
Gene Symbol: Ttc39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145948.1 | NP_001139420.1 | 15, 16 | 1444 | ||
NM_153392.2 | NP_700441.2 | 15, 16 | 1453 | |||
GenBank mRNA | AK048659.1 | - | 15, 16 | 1454 | ||
AK076641.1 | - | 15, 16 | 1407 | |||
AK076720.1 | - | 15, 16 | 1446 | |||
AK076772.1 | - | 15, 16 | 1408 | |||
AK172951.1 | - | 15, 16 | 1376 | |||
BC023883.1 | - | 15, 16 | 1452 | 5150841 | ||
BC026660.1 | - | 15, 16 | 1394 | 4988900 |
