siRNA Details
| Gene Symbol: Epha10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256432.1 | NP_001243361.1 | 1, 2 | 197 | ||
| NM_177671.5 | NP_808339.2 | 1, 2 | 197 | |||
| GenBank mRNA | AK039700.1 | - | 1, 2 | 160 | ||
| AK143168.1 | - | 1082 | ||||
| BC116991.1 | - | 1, 2 | 127 | 40126310 | ||
| BC119084.1 | - | 1, 2 | 127 | 8733833 | ||

Epha10
Genomic Map