siRNA Details
Gene Symbol: Phf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172705.2 | NP_766293.2 | 4 | 1144 | ||
GenBank mRNA | AK077052.1 | - | 2 | 658 | ||
AK132066.1 | - | 4 | 1149 | |||
AY069976.2 | - | 4 | 1069 | |||
BC020286.1 | - | 2 | 193 | |||
BC029632.1 | - | 4 | 1144 | 3500487 | ||
BC059282.1 | - | 4 | 1144 | 3500487 |
